(1S,2S)-(+)-1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S,S)-Jacobsen Ligand, (S,S)-(+)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine

07-0317-1G 07-0317-5G
100206-224EA 48.12 USD
100206-224 100195-964
(1S,2S)-(+)-1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene) ≥98%
(1S,2S)-(+)-1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)

Jacobsen Ligand

Formula: C₃₆H₅₄N₂O₂
Melting Pt: 205…207 °C
MDL Number: MFCD00191801
CAS Number: 135616-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR