(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol ≥98%

Supplier: STREM CHEMICALS INC MS

07-0201-5G 07-0201-1G
100200-624EA 186.83 USD
100200-624 100199-716
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol ≥98%
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol

CATHy Catalyst

Formula: C₉H₁₁NO
Melting Pt: 117…121 °C
MDL Number: MFCD00216656
CAS Number: 136030-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR