(4R,5S)-(+)-4-i-Propyl-5-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)-2-oxazolidinone ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

07-0099-500MG 07-0099-100MG
101433-058EA 594.46 USD
101433-058 101433-056
(4R,5S)-(+)-4-i-Propyl-5-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)-2-oxazolidinone ≥99%
(4R,5S)-(+)-4-i-Propyl-5-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)-2-oxazolidinone
Melting Pt: 96…99 °C
MDL Number: MFCD09834835
CAS Number: 1432793-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR