2,2'-Dibromo-1,1'-binaphthyl ≥96%

Supplier: STREM CHEMICALS INC MS
Warning

06-0450-1G 06-0450-5G
100199-710EA 117.48 USD
100199-710 100199-712
2,2'-Dibromo-1,1'-binaphthyl ≥96%
2,2'-Dibromo-1,1'-binaphthyl
Formula: C₂₀H₁₂Br₂
Melting Pt: 178…181 °C
MDL Number: MFCD00188005
CAS Number: 74866-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR