2-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 2-Ethoxycarbonylphenylboronic acid pinacol ester, 2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, Ethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, Ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

05-0770-5G 05-0770-1G
100204-774EA 560.49 USD
100204-774 100204-772
2-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥97%
2-(Ethoxycarbonyl)phenylboronic acid pinacol ester

Boronic Acid and Ester

Formula: C₁₅H₂₁BO₄
Melting Pt: 55…58 °C
MDL Number: MFCD03453660
CAS Number: 269409-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR