Human Recombinant Serum Amyloid P-Component
Supplier: Bon Opus Biosciences
C510-10UG C510-1MG C510-500UG C510-50UG
102862-646EA 90.46 USD
102862-646 102862-648 102862-650 102862-652
Human Recombinant Serum Amyloid P-Component
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Serum Amyloid P Component is produced by our Mammalian expression system and the target gene encoding His20-Val223 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.