Nickel 50% slurry in water, sponge for use as a catalyst

Supplier: STREM CHEMICALS INC MS
Danger

28-1890-100G 28-1890-500G
100206-558EA 264.68 USD
100206-558 100201-572
Nickel 50% slurry in water, sponge for use as a catalyst
Nickel

Stability: air sensitive
Physical Characteristics: surface area 80-100 m2/g

Formula: Ni
Density: 8.90 (as nickel metal)
MDL Number: MFCD00011137
CAS Number: 7440-02-0
UN: 1378
ADR: 4.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR