IPI-145 ≥99% (by HPLC)

Supplier: Enzo Life Sciences

CHM223-0010 CHM223-0025 CHM223-0050 CHM223-0100 CHM223-0200 CHM223-0500
76203-194EA 153 USD
76203-194 76203-196 76203-198 76203-200 76203-202 76203-204 76203-206
IPI-145 ≥99% (by HPLC)
IPI-145

Duvelisib is an orally bioavailable and highly selective inhibitor of PI3K-δ and PI3K-γ with IC50 values of 0.36 nM and 19.6 nM, respectively.


  • Highly selective inhibitor of PI3K-δ and PI3K-γ
  • White solid
  • Soluble in DMSO.

Formula: C₂₂H₁₇ClN₆O
Storage Temperature: Freezer
MDL Number: MFCD15144635
CAS Number: 1201438-56-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d17ca0f52c9d6e250a802e78814560ba61227772&vid=4faf7876b5446b196d76c07402917d35d0f6981a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR