Di[tris(hydroxymethyl)aminomethane] carbonate (TRIS carbonate) ≥99%

Supplier: CHEM IMPEX INTL INC MS

Synonyms: 2-Amino-2-hydroxymethyl-1,3-propanediol carbonate

01198-100G 01198-250G 01198-1KG 01198-25G
102555-486EA 105.97 USD
102555-486 102555-488 102588-072 102615-778
Di[tris(hydroxymethyl)aminomethane] carbonate (TRIS carbonate) ≥99%
Di[tris(hydroxymethyl)aminomethane] carbonate (TRIS carbonate)
Formula: (C₄H₁₁NO₃)₂·CH₂O₃
MW: 304.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038952
CAS Number: 68123-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR