N-Acetyl-DL-leucine 99%
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
N-Acetyl-DL-leucine, Ac-DL-Leu-OH, 2-Acetamido-4-methylpentanoic acid
06647-25G
06647-1KG
06647-100G
06647-250G
102576-762EA
32.2
USD
102576-762
102576-764
102620-094
102620-096
N-Acetyl-DL-leucine 99%
N-Acetyl-DL-leucine
Synonym(s): Ac-DL-Leu-OH, (RS)-Ac-2-amino-4-methylpentanoic acid
Formula:
C₈H₁₅NO₃ MW: 173.2 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00026498 CAS Number: 99-15-0 |
Specification Test Results
Purity | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...