Triphosgene ≥99%, white to off white crystals
Supplier: CHEM IMPEX INTL INC MS
|
Danger
|
Synonyms:
Bis(trichloromethyl) carbonate
07205-5KG
07205-1KG
07205-25G
07205-100G
07205-250G
102562-782EA
2125.05
USD
102562-782
102607-466
102620-388
102620-390
102620-392
Triphosgene ≥99%, white to off white crystals
Triphosgene
Synonym: Bis(trichloromethyl) carbonate
Formula:
C₃Cl₆O₃ MW: 296.75 g/mol Boiling Pt: 203…206 °C Melting Pt: 76…82 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00062848 CAS Number: 32315-10-9 UN: 2928 ADR: 8,II |
Specification Test Results
Purity | ≥99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...