DuPont™ Tychem® 5000 Coveralls with Mandarin Collar
Supplier: DuPont
C3184TTN2X000600 C3184TTN3X000600 C3184TTN4X000600 C3184TTNLG000600 C3184TTNMD000600 C3184TTNSM000600 C3184TTNXL000600
76187-722CS 1383.24 USD
76187-722 76187-724 76187-726 76187-728 76187-730 76187-732 76187-734
DuPont™ Tychem® 5000 Coveralls with Mandarin Collar
Disposable Garments
Tychem® 5000 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate.
  • Mandarin collar
  • Attached dual-layer gloves
  • Multi-layer laminate interior gloves
  • Neoprene exterior gloves
  • Attached socks with outer boot flaps
  • Double storm flap with hook & loop closure
  • Taped seams
Tychem® 5000 fabric is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. Typical Applications: chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness. Commonly used in domestic preparedness for situations where the potential to exposure to chemicals exist. These coveralls are tan.
Tychem® CPF3 HD garments were designed with extensive input from first responders to ensure that HAZMAT, law enforcement, military and hospital personnel get the protection they need from chemical warfare agents (including Sarin, Mustard, and Lewisite) and toxic industrial chemicals (TIC's). Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Mandarin collar is designed to fit tightly around hooded PAPR. Larger zipper pull is easier to grasp for easier donning and doffing. Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper. Attached internal gloves are composed of five layer laminate film for a broad range of chemical protection. Neoprene outer gloves deliver high tactility and dexterity to handle tools or weapons. Integrated socks are composed of the same garment material. Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion.
Certifications: Thickness meets ASTM D1777.