Anti-Histiocytoma Marker Mouse Monoclonal Antibody (CF647) [clone: D11]
Supplier: Biotium
BNC470348-100 BNC470348-500
75950-538EA 205.77 USD
75950-538 75950-540
Anti-Histiocytoma Marker Mouse Monoclonal Antibody (CF647) [clone: D11]
Antibodies
In Western blotting, this antibody detects an antigen of 125 kDa in human liver and 135 kDa in tumors of histiocytic origin. Comparative study of this MAb and a standard CD68 MAb showed that their antigens are different. Its antigen in all macrophage types studied is located on the plasma membrane and within cytoplasmic structures including lysosomes. This MAb shows a restricted reactivity to cells of the monocyte/macrophage system. It specifically reacts with blood monocytes and stains resident macrophages in a wide variety of human tissues. This MAb does not stain antigen-presenting cells,More Product Information