Orbital Shakers, Bellco Glass
Supplier: Bellco Glass
7744-02020 7744-01010
100486-024CS 1964.38 USD
100486-024 100486-022
Orbital Shakers, Bellco Glass
Shakers and Mixers Orbital Shakers
Standard orbital shaker ideal for hybridization and gel agitation.
  • It has rubber lined tray with 1/2" offset providing a circular orbit
  • Peg and grommet platform design for easy setup