TMAO Medium, HiMedia Laboratories
Supplier: HiMedia
M1159-500G M1159-2.5KG M1159-5KG
95020-600EA 245.02 USD
95020-600 89223-158 89223-160
TMAO Medium, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
TMAO (Trimethylamine-N-Oxide Medium) medium is used for the cultivation and differentiation of Campylobacter species from foods, except C. jejuni and C. coli in accordance with APHA.