Tartoff - Hobbs (Terrific) Broth, HiMedia Laboratories
Supplier: HiMedia
GM1250-2.5KG
GM1250-500G
GM1250-5KG
75804-052EA
844.02
USD
75804-052
75804-054
75804-056
Tartoff - Hobbs (Terrific) Broth, HiMedia Laboratories
Media for Microbiology
Dehydrated Media for Microbiology
Tartoff-Hobbs (Terrific) broth is used for the cultivation of recombinant strains of Escherichia coli.
- Cream to light yellow coloured
- Granular medium
- Reaction of 4.76% w/v aqueous solution at 25 °C
Caution: Store below 30 °C in tightly closed container and the prepared medium at 2 to 8 °C.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)