Glycogen (from oysters), Ultrapure

Supplier: Thermo Fisher Scientific

J16445-14 J16445-06 J16445-22
101171-658EA 468.55 USD
101171-658 101169-902 103728-102
Glycogen (from oysters), Ultrapure
Glycogen
Formula: C₂₄H₄₂O₂₁
MW: 666.579 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081547
CAS Number: 9005-79-2
EINECS: 232-683-8

Order Now

Specification Test Results

Form Light tan powder
Identification (FTIR) Conforms
Loss on Drying (110°C, 4 hrs.) ≤ 15%
Optical Rotation (c=0.2, H2O, dry basis) +170° to 200°
Solution Test (0.25%, 1M NaCl) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR