Sulfuric acid 50% (v/v) 18 N

Supplier: AQUA SOLUTIONS MS
Danger

9000-1L 9000-20L 9000-500ML 9000-4L 9000-500MLG
101223-108EA 48.03 USD
101223-108 101225-186 101226-548 101225-318 103193-862
Sulfuric acid 50% (v/v) 18 N
Sulfuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 100 °C (212 °F)
Density: 1.42 g/cm³ (11.85 lbs/gal.) (20 °C)
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 2796
ADR: ,II

Order Now

Specification Test Results

Assay 50%
Normality 18
Appearance Clear Liquid
Vapor Pressure (20 °C) 23 hPa (17 mm Hg)
Density (20 °C) 1.48231 - 1.50189
Odor Odorless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR