Tiger Inox, Combo Wheels, Weiler®
Supplier: Safety & Industrial Supplies
804-58114 804-58115
76184-300EA 84.34 USD
76184-300 76184-302
Tiger Inox, Combo Wheels, Weiler®
Abrasives
Delivers true flexibility when switching between cutting and grinding for stainless steel applications.
  • Contaminate-free white aluminum oxide wheel
  • Fiberglass body
  • Resin bonded
  • Long lasting and fast cutting
  • Use with right angle grinders
Use with high-speed electric rotary tools or air-powered pencil die grinders.