Compound 34 ≥98%
89147-846EA
126
USD
89147-846
89147-848
Compound 34 ≥98%
Compound 34
γ-Secretase inhibitor
Cell permeable, highly potent inhibitor of γ-secretase (IC50=0.06nM).
Formula:
C₃₁H₂₄F₃N₃O₃ |
CAS Number:
564462-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...