Anti-GT1a Mouse Monoclonal Antibody [clone: GMR11]

Supplier: TCI
A2702-VIAL
TCA2702-VIALEA 1041 USD
TCA2702-VIAL
Anti-GT1a Mouse Monoclonal Antibody [clone: GMR11]
Antibodies
Purified mouse monoclonal antibody/IgM

Type: Primary
Antigen: GT1a
Clonality: Monoclonal
Clone: GMR11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR