Weiler® Wide-Face Standard Twist Knot Wire Wheel, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-09480 804-09430 804-09540 804-09490 804-09160
300042-294EA 231.68 USD
300042-294 300041-582 300041-706 300010-141 300042-864
Weiler® Wide-Face Standard Twist Knot Wire Wheel, ORS Nasco
Brushes
Rugged knot-type construction provides aggressive, high-impact action for severe applications.
  • Heavy-duty brushing action with some flexibility
  • Double row of knotted wire for a wider brush face
  • Can be used with bench/pedastal grinders
Designed for coating removal, heavy deburring, removal of excess molded rubber, heavy corrosion, rust, paint, oxides, and scale, and surface roughening and preparation.