Aurion Biotinylated Albumin/Gold, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
25290
25291
25299
100487-674EA
592.45
USD
100487-674
100487-684
100487-670
100487-672
100487-676
100487-680
100487-682
100487-686
100487-688
100487-678
Aurion Biotinylated Albumin/Gold, Electron Microscopy Sciences
Protein Linking and Labeling Systems
Biotinylated albumin conjugates contain equimolar amounts of conjugated proteins.
- Gold particle sizes of 0.8, 10 ,15 and 25 nm
- Monodisperse particle population
- Minimal size variation and overlap