L(+)-Histidine ≥99%, white crystalline powder cell culture reagent

Supplier: 0000012207

0219468490 0219468491
76177-732EA 847.18 USD
76177-732 76177-734
L(+)-Histidine ≥99%, white crystalline powder cell culture reagent
L(+)-Histidine

Storage: Store at Room Temperature (15-30 °C.)
L-histidine is one of the three amino acids with a basic side chain, and is very hydrophilic in character. It contains an imidazole group in the side chain.
L-Histidine has been used to study cultures of the human T-lymphoblastic leukemia cell line MOLT-4 to study modulation of apoptosis. It is used in treatment of rheumatoid arthritis, allergic diseases, ulcers & anemia.
Precursor of histamine by action of histidine decarboxylase.


  • Cell Culture Reagent Grade
  • Presentation: White Crystalline Powder
  • Water soluble 50 mg/mL.

Formula: C₆H₉N₃O₂
Melting Pt: 282 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00064315
CAS Number: 71-00-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=badfdb36aac0611e5823a7d05e6cba944f6650fa&vid=9e9f9d881a5b3314c7f342312e77e13493901200&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
pH (1% aq soln) 7.0-8.5
Loss on Drying ≤0.5%
Optical Rotation (c=10, 4N HCl) +11.0° to +13.5°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR