Weiler® Roughneck® Stringer Bead Wheel, ORS Nasco
Supplier: Safety & Industrial Supplies
804-08776 804-09500 804-09200 804-08916 804-08766 804-13131P 804-13128 804-08756 804-13131 804-13138P 804-09400 804-09000 804-09000P 804-13232 804-13238
300010-140EA 46.88 USD
300010-140 300010-142 300043-274 300043-406 300044-220 300045-200 300045-314 300001-011 300001-014 300045-202 300001-012 300001-013 300043-114 300085-300 300085-302
Weiler® Roughneck® Stringer Bead Wheel, ORS Nasco
Designed to remove large burrs and heavy contamination, including oxides, rust, paint, and scale.
Use with right angle grinders and straight grinders.