Mineral oil light, liquid for molecular biology
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Mineral Oil is a colorless, odorless mixture of alkanes from a mineral source. It is often used as an inert overlay to minimize evaporation in molecular biology applications, including PCR, but has many other applications on other research fields.
- Refractive Index: n20/D 1.467 (lit.)
Mineral oil is suitable for use as an overlay to control evaporation and cross contamination of samples in a variety of molecular biology applications.
Boiling Pt:
218...643 °C (1013 hPa) (lit.) Density: 0.81…0.894 (15 °C) (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00131611 CAS Number: 8042-47-5 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...