Tweezers WA1 Handle with 16-ESD Tips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
TIP-16-ESD 10004-E16-SM
103301-842EA 12.5 USD
103301-842 103302-558
Tweezers WA1 Handle with 16-ESD Tips, Electron Microscopy Sciences
Forceps
Swiss made precision tweezers from Dumont.
  • Hand made of the highest quality steels and alloys
  • Perfectly finished to suit your application
These tweezers have an anti-magnetic WA1 handle with replaceable 16-ESD tips. Tips attach with screws. Length is 130 mm.
Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox.