Weiler® Crimped Wire Cup Brush, ORS Nasco
Supplier: Safety & Industrial Supplies
804-13188 804-14066 804-14316 804-14256 804-13240 804-14036 804-14616 804-13231 804-13245P 804-14606 804-13245 804-14216 804-14126 804-13181 804-14026 804-14076 804-14166 804-14206 804-14016 804-13243 804-13244
300010-169EA 96.63 USD
300010-169 300010-175 300042-458 300042-938 300044-588 300000-988 300043-354 300044-586 300044-700 300042-940 300000-989 300042-936 300043-352 300000-990 300010-174 300010-176 300042-566 300042-934 300043-350 300044-578 300044-580 300044-584 300044-590 300044-592 300044-594
Weiler® Crimped Wire Cup Brush, ORS Nasco
Brushes
These crimped brushes are designed for use with right-angled grinders to remove rust, paint, oxides, and scale.
  • For light to medium duty, general-purpose applications
  • Very flexible brushing action with consistent performance
  • Features a threaded nut for use on angle grinders
Applications include cleaning, removal, roughening, and deburring.