Weiler® Crimped Wire Solid End Brushes, ORS Nasco
Supplier: Safety & Industrial Supplies
804-10003 804-10012 804-10014 804-10017 804-10023 804-10009 804-10128 804-10020 804-10006 804-10158 804-10019 804-10021 804-10022 804-10001 804-10007 804-10010 804-10011 804-10013 804-10018 804-10024 804-10005 804-10002 804-10004 804-10015 804-10016 804-10008
300010-144EA 15 USD
300010-144 300010-148 300010-150 300010-151 300010-155 300046-678 300046-688 300047-704 300047-702 300048-442 300091-882 300010-153 300010-154 300010-143 300010-145 300010-146 300010-147 300010-149 300010-152 300046-680 300047-700 300048-434 300048-436 300048-438 300048-440 300087-840
Weiler® Crimped Wire Solid End Brushes, ORS Nasco
Brushes
Crimped brushes are ideal for general-purpose and light duty applications to remove rust, paint, oxides, and scale.
  • Light-duty cleaning in corners and hard to reach areas
  • Solid end construction
  • Very flexible brushing action
Use with power drills, drill presses, die grinders, and right-angle air die grinders.