CUNO™ CTG-Klean System Filter Pack with MicroKlean™ RB Series Filter Cartridge
Supplier: 3M Healthcare
7010413591 7010409948
76028-905CS 1324.12 USD
76028-905 76029-257 76031-771
CUNO™ CTG-Klean System Filter Pack with MicroKlean™ RB Series Filter Cartridge
Filters Liquid Process Filters
CUNO™ CTG-Klean System Filter Pack with Micro-Klean™ RB Series Filter Cartridge is part of a totally enclosed system that uses a separate pressure vessel and disposable filter pack to isolate the product to be filtered from the housing. It reduces the time, labor and amount of cleaning solvents required during filter change-out and cleaning of conventional housings.
  • Graded porosity filter cartridge for low pressure drop and long service life
  • Rigid, resin-bonded filter cartridge structure produces no by-pass or unloading with high pressure drops or surges
  • Broad range of micron ratings offered, from 1 to 150 μm, to meet a wide range of filtration requirements
  • Filter pack isolates the product to be filtered from the housing
Advancing Depth Filtration.
Micro-Klean™ RB Series filter’s rigid construction is designed to reduce by-pass and avoid the unloading characteristics of soft and easily deformable meltblown and stringwound filter cartridge designs. Micro-Klean™ RB series filter cartridges are the product of continuous refinement of our manufacturing and fiber technologies. Available in both grooved and ungrooved versions, they are ideal for a wide variety of applications. The grooving of the outer surface significantly increases the filter’s effective surface area, and increases the contaminant holding capacity. The ungrooved version of the Micro-Klean RB series cartridge is effective for the reduction of gels and other deformable contaminants. To provide compatibility with a wide range of process fluids, Micro-Klean RB series cartridges are available in several combinations of fiber type and resin.
Applications include Paints, Inks, Emulsions, Adhesives, Resins, Organic Solvents, Coolants, Lube Oils, Various Chemicals, Pesticides and Fertilizers.
Note: Micro-Klean™ RB series cartridges are not designed for, and should not be used in Food and Beverage and Pharmaceutical applications.