3M™ Zeta Plus™ Single Use Filter Capsule with ZB Series Media
Supplier: 3M Healthcare
7010381424 7100049898 7010339021 7010339022 7010301514 7010381431 7010381432 7000144882 7000144883 7010381437 7010409939 7010381420 7010340353 7100085520 7010301513 7100030469 7100030470 7010339026 7010301515 7100049903 7100026714 7100021689 7000144884 7000144885 E0340FSA120ZB05A E0340FSA120ZB08A E0340FSA120ZB10A E0340FSA60ZB05A E0340FSA90ZB05A E0340FSA90ZB08A E1020FSA120ZB E1020FSA120ZB05A E1020FSA120ZB08A E1020FSA120ZB10A E1020FSA60ZB05A E1020FSA90ZB05A E1020FSA90ZB08A E0170FSA120ZB05A E0170FSA120ZB08A E0170FSA120ZB10A E0170FSA30ZB E0170FSA60ZB05A E0170FSA90ZB05A E0170FSA90ZB08A
76030-756CS 651.01 USD
76030-756 76030-762 76030-772 76030-782 76030-792 76030-808 76030-818 76030-820 76030-822 76030-838 76030-844 76033-228 76033-236 76033-240 76033-250 76033-262 76033-268 76033-286 76033-292 76033-300 76033-302 76046-972 76046-974 76046-976 77675-214 77675-182 77675-170 77675-218 77675-186 77675-216 77675-180 77675-146 77675-158 77675-196 77675-140 77675-130 77675-176 77675-154 77675-144 77675-134 77675-184 77675-212 77675-132 77675-112
3M™ Zeta Plus™ Single Use Filter Capsule with ZB Series Media
Filters Capsule Filters
Zeta Plus™ ZB series filter media is an advanced depth filter media designed to provide clarification of bioprocess, biological and pharmaceutical fluids.
  • Broad pH range of strong, positively charge-modifed depth flter media
  • Economical, effcient reduction of negatively charged contaminants such as cell debris, colloidal material, endotoxin, DNA, RNA, and bacteria
  • Pre-extracted inorganic flter aid
  • Eases validation and regulatory submissions by providing vital documentation and data traceability
Zeta Plus ZB series filter media is composed of pre-extracted inorganic filter aid, cellulose and a resin system that imparts a strong positive charge to the filter matrix, aiding its adsorption of negatively charged contaminants.
Certifications: FDA Drug Master File and USP Class V Biological Safety.