3M™ Aqua-Pure™ Whole House Water Filtration Systems, AP900 Series
Supplier: 3M Healthcare
7100007699 7100039277
76028-696EA 957.61 USD
76028-696 76028-698
3M™ Aqua-Pure™ Whole House Water Filtration Systems, AP900 Series
Filters Filter Cartridges
3M™ Aqua–Pure™ whole house sanitary quick change water filter system AP902 contains high performing, multitasking filters that help reduce contaminants such as sediment, chlorine taste and odor and now even scale. These systems are easy to install, maintain and change out.
  • Sanitary Quick Change (SQC) design allows for fast and easy cartridge change–outs without the need for a filter wrench
  • Up to 20 gallons per minute (GPM) flow rate meets the water pressure demands of most homes
  • Manufactured from FDA CFR–21 compliant materials that are food and drinking water safe
  • 304 Stainless Steel Head provides 'sweat–in–place' installation, corrosion resistance, and long life
  • Whole house water filtration helps reduce sediment build–up and increase the life of hot water heaters and appliances
Features include: 1" NPT Stainless Steel Inlet and outlet plumbing connections, Peak flow rate is up to 20 gpm/75.7 lpm, System maximum operating pressure of 125 psi (862 kPa) and operating temperature of 100 °F/37.8 °C, The AP902 system incorporates a non–woven pleat media that reduces sediment down to 5 µm nominal, The AP903 incorporates carbon block filtration with a non–woven pleat media to reduce sediment down, The AP904 system incorporates scale inhibition media, carbon block filtration and a non–woven pleat media to reduce sediment down to 5 µm nominal, chlorine taste and odor, as well as scale build – up, System materials are FDA CFR–21 compliant and are food and drinking water safe, Cartridges are sanitary in design, requiring no contact with the filter media during cartridge change–out.
The scale inhibition feature uses polyphosphates in minute quantities to bind calcium and magnesium hardness minerals, preventing them from precipitating out and building up on metallic surfaces which can reduce flow rates throughout the homes plumbing system. 1" stainless steel inlet and outlet connections provide easy installation and high flow rates up to 20 gallons per minute (GPM) / 75.7 liters per minute (LPM) to meet the needs of residential and light commercial applications. The innovative Sanitary Quick Change (SQC) design, an industry first for this application, allows for fast and easy cartridge change–outs without having to contact the filter media or the need for a filter wrench.
The AP902 Series is manufactured with a head assembly that includes an integral mounting bracket. The 304 stainless steel head allows for 'sweat in place' installation and is a corrosion resistant material that provides long service life and won't affect the taste of drinking water. The stainless steel head comes with a 25 year warranty for worry–free ownership. The AP900 Series line also includes an electronic countdown timer refrigerator magnet to remind the owner when to change the cartridge.