3M™ High Flow Series Filter Housing
Supplier: 3M Healthcare
7000001847 7000125552
76028-844CS 1730.37 USD
76028-844 76029-156
3M™ High Flow Series Filter Housing
Filters Filter Cartridges
3M™ High Flow Series Filter Housings are specifically designed to deliver all of the filtration benefits in a compact footprint. Housings are available in standard designs, as well as customizable configurations to suit your specific needs. All standard 3M high flow series filter housings are designed, manufactured, tested, and code stamped in accordance with ASME Section VIII, Division 1.
  • 3M™ High Flow Series Filter Housings are 33 to 50% smaller than competitively sized housings for a given flow rate
  • Robust cartridge center–post design eliminates bulky support plates providing easy access to housing internals
  • Hinged cover (horizontal units) or user–friendly cover lifting device (vertical units) for easy element change–outs
  • Manufactured from carbon steel, 304 or 316L stainless steel for effective corrosion protection
  • ASME Code design meets high quality and performance standards
3M High Flow Series Filter Housings are 33 to 50% smaller than competitively sized housings for a given flow rate
Our housings are designed with features that make them easy to use and streamline processes. Spend less time cleaning thanks to removable cartridge posts that lift out for easy cleaning, which helps maintain cleaner systems and longer product life. To help ensure against bypass, leaks, and maintain system pressure, shouldered seal plates fit solidly into the cartridge core for a positive cartridge seal. Housing drains for both sludge and clean chambers feature drain couplings for easy draining. An air vent facilitates effective venting before start up or draining.
Fewer filters and a user–friendly housing design means faster change–outs than competitively sized systems.
Fewer filters and a user–friendly housing design means faster change–outs than competitively sized systems. Our high flow series filter housings are available in a variety of sizes to accommodate from 1 to 7 filter elements in both 40 and 60" lengths. Choose the horizontal option to maximize ease of operation, or the vertical to minimize the system's footprint.