3M™ LifeASSURE™ PFS Series Filter Cartridge
Supplier: 3M Healthcare
7000050880 7000029190 7000050553 7000029191 7000050259 7000029206
76033-614CS 12330.7 USD
76033-614 76034-057 76034-059 76034-061 76034-063 76034-065
3M™ LifeASSURE™ PFS Series Filter Cartridge
Filters Filter Cartridges
LifeASSURE™ PFS Series PTFE filter cartridges combine flow-enhancing construction with a liquid‐validated, bacteria retentive, hydrophobic PTFE filter membrane, providing excellent air flow rates using small assemblies. Filters are used for applications requiring 0.2 µm sterilization of air, gas, solvents and chemicals in life science industries.
  • Flow-enhancing construction allows air flow with a relatively low pressure drop, resulting in a smaller vent filter assemblies
  • Hydrophobic polytetrafluoroethylene (PTFE) membrane helps prevent pore blockage in vent applications, extending filter lifespan
  • Liquid validation of B. diminuta retention helps provide reliable sterilizing performance in wet or dry conditions
  • 100% forward flow integrity tested prior to release
Validated PTFE membrane.
Our LifeASSURE™ PFS series PTFE Filter Cartridges use a hydrophobic polytetrafluoroethylene (PTFE) membrane, which helps minimize pore blockage from moisture in vent applications. Our filters are validated by a liquid bacterial challenge for complete retention based on the ASTM standard, which provides a high level of reliable sterilizing performance.
High air flow construction
Our LifeASSURE™ PFS Series filter cartridges are constructed to achieve high air flow at a given pressure drop. The PTFE membrane is pleated with polypropylene nonwoven flow-enhancing layers during construction to achieve optimal flow rate performance at low differential pressures.
We manufacture LifeASSURE™ PFS Series filter cartridges using all polypropylene structural components (end caps, adapters, media support, inner core and outer cage). The 3M Purification Inc. Quality Management System is approved by an accredited registering body to the ISO 9001:2008 and ISO 13485:2003 Quality Management Systems, and the cartridges are 100% forward flow integrity tested prior to shipment. All LifeASSURE PFS Series filter cartridges are shipped with a Certificate of Quality affirming compliance with manufacturing quality specifications. Supporting Drug Master File (DMF) documentation is on file with the United States Food and Drug Administration (FDA).