3M™ LifeASSURE™ PNA Series Filter Cartridge
Supplier: 3M Healthcare
7000029544 7010381202 7000051404 7000001853 7000051419 7000051403 7000029540
76032-757CS 8743.65 USD
76032-757 76032-759 76032-761 76034-163 76034-165 76034-167 76034-169
3M™ LifeASSURE™ PNA Series Filter Cartridge
Filters Filter Cartridges
The 3M™ LifeASSURE™ PNA Series Filter Cartridge offers robust 0.20 and 0.45 µm absolute-rated filtration by combining three leading-edge processes – hydrophilic asymmetric polyethersulfone membrane construction, Advanced Pleating Technology and an upstream/downstream support design. This series is ideally suited for bio-burden applications.