3M™ LifeASSURE™ BLA Series Filter Cartridge
Supplier: 3M Healthcare
7000050301 7010313741 7000050304 7100010839 7000050306 7010242349 7100039000 7100007550 7000050311 7000050235 7010379300 7000050601 7010338075 7100025231 7010301318 7000050464 7010339097 7000050606 7010381115 7000029308 7000050609
76029-777CS 2455.96 USD
76029-777 76029-779 76029-781 76029-783 76029-785 76029-789 76029-791 76029-793 76029-795 76029-797 76029-799 76029-801 76029-803 76029-805 76029-807 76029-811 76029-813 76029-815 76029-817 76029-819 76029-823
3M™ LifeASSURE™ BLA Series Filter Cartridge
Filters Filter Cartridges
LifeASSURE™ BLA Series Filter Cartridges are ideal for pre-filtration and clarification for food and beverage applications. BLA series prefilters help protect and extend final filter service life, decrease change-outs, and reduce effluent microorganism content.
  • Multi-zone, microporous Nylon 6,6 membrane filtration, available in 0.2 to 0.8 micron range for protection of final membrane filter cartridges
  • Advanced Pleat Technology (APT) construction provides extremely high surface area for higher throughputs
  • 100% Integrity tested in manufacturing and in situ for proper installation and operation
  • Made with FDA CFR-21 compliant materials
LifeASSURE™ BLA series filter cartridges offer high performance as either final filters for spoilage organism removal, or as a pre-filter. The outer cage, inner core, end-cap adapters and media support layers of these filter cartridges are made of PP.
In prefiltration, they are effective in prolonging the service life of LifeASSURE™ BA, BNA and BDA final filters and help protect other downstream membrane filters. In manufacturing LifeASSURE BLA series filter cartridges we encompass two leading-edge processes, multi-zone microporous membrane manufacture and our Advanced Pleat Technology (APT) construction. This technology maximizes the useful surface area of the filter and maintains proper flow paths between media pleats. By employing more effective surface area, LifeASSURE BLA series filter cartridges provide low pressure drops and a long service life. Our filters are ideally suited to meet a wide range of filtration and clarification requirements in the beverage industry. As a result of their high-performance and long service life, LifeASSURE BLA series filter cartridges help lower related operational costs.
Significant micro-organism reduction is the key.
For effective filtration and clarification of beverages, micro-organism reduction is a crucial outcome needed from economical, reliable filter systems. In tests with Brevundimonas diminuta, LifeASSURE BLA™ series 020 grade filters exhibited an average log reduction value (LRV)* of 7.3 and the LifeASSURE BLA series 045 grade filters exhibited an average LRV of 3.5.
* LRV=log10 (number of organisms in/number of organisms out).
Applications include wine, beer, bottled water, soft drinks, syrups, food grade dyes and flavors, preservative solutions, blending and wash water and distilled spirits.
Certifications: Recommended filter cartridge change-out differential pressure: 2.4 bar (Recommended filter cartridge change-out differential pressure: 35 psid)