3M™ LifeASSURE™ IMC Series Filter Cartridge
Supplier: 3M Healthcare
7010296366 7100169695 7010293009
76032-433CS 4346.57 USD
76032-433 76032-435 76032-441
3M™ LifeASSURE™ IMC Series Filter Cartridge
Filters Filter Cartridges
LifeASSURE™ IMC Series Filter Cartridge is a high efficiency naturally hydrophilic Nylon 6,6 filter elements designed to meet the exacting requirements of deionized (DI) water, critical parts cleaning, and industrial chemical applications.
  • No IPA pre-wetting and system flushing required
  • Enhanced contaminant reduction capacity
  • Excellent downstream cleanliness out of the package
  • Economic alternative to PTFE filter cartridges
Utilizing Advanced Pleat Technology (APT), the LifeASSURE™ IMC filter cartridge provides enhanced flow characterizes with minimal pressure drop.
Applications include DI water, critical parts cleaning, chemicals.
This filter is available in single and multiple length cartridges with 0.1, 0.2, 0.45, 0.65 and 0.80 µm ratings along with various end modifications. The naturally hydrophilic Nylon 6,6 membrane provides low extractable, long filter life and enhanced contaminant reduction. LifeASSURE™ IMC filter cartridges are ideally suited for DI water, critical parts cleaning, and other chemical applications where high efficiency contaminant reductions at 0.1, 0.2, 0.45, 0.65 and 0.80 μm is required.