Type 29 Tiger Paw Angled Flap Discs, Weiler®
Supplier: Safety & Industrial Supplies
804-51124 804-51121 804-51126 804-51119 804-51120 804-51125
300053-356CS 107.79 USD
300053-356 300053-354 300053-360 300053-350 300053-352 300053-358
Type 29 Tiger Paw Angled Flap Discs, Weiler®
Abrasives
  • 100% zirconia alumina grain is fast cutting with aggressive action and long life, resulting in fewer changeovers and increased productivity
  • Engineered poly/cotton cloth features excellent grain retention for maximum disc life