RS Medium Duty Single Stage Regulators, Western Enterprises
Supplier: Western Enterprises
312-RS-2-4 312-RS-9-4 312-RS-9-3 312-RS-7-3
300066-972EA 304.2 USD
300066-972 300066-974 300066-980 300066-982
RS Medium Duty Single Stage Regulators, Western Enterprises
Welding Supplies
The 450 psig models have KEL-F® seat to accommodate high pressure applications
  • Dual inlet filter system extends regulator life
  • Machined brass body
  • Neoprene diaphragm
  • Nickel chrome plated zinc bonnet
Hydrogen and methane models are equipped with pipe away relief valves.
Certifications: UL Listed.