Regulator Bushing Adaptors, Western Enterprises
Supplier: Safety & Industrial Supplies
312-CV-30R 312-CV-31L 312-WE-61
300007-971EA 17.81 USD
300007-971 300007-972 300008-024
Regulator Bushing Adaptors, Western Enterprises
Welding Supplies
  • Easily mounts on outlet bushing on a regulator having a B-Size outlet
  • This model has the same flow capacity as the torch - type (1200 SCFH at 50 psig) when installed on a regulator
  • Check valve is easily seen and is less subject to contamination, abuse, and damage
Certifications: Meets or Exceeds CGA and OSHA Requirements.; UL Listed.