Pimamicin ~99%

Supplier: SELLECK CHEMICALS LLC MS

Synonyms: Natamycin

S1517-50MG S1517-200MG
101760-838EA 192.31 USD
101760-838 101760-840 101760-842
Pimamicin ~99%
Pimaricin

Pimaricin ( synonym Natamycin), a natural and versatile anti-fungal agent during fermentation by the bacterium Streptomyces natalensis, commonly found in soil, with little to no flavour interference.


  • DMSO Solubility of 7 mg/mL (10.51 mM)


Natamycin binds specifically to ergosterol present in model membranes. Natamycin does not change the permeability of the yeast plasma membrane under conditions that growth is blocked. Natamycin perturbs the cellular vacuole morphology, causing the formation of many more small vacuolar structures in yeast cells. Natamycin inhibits the priming stage of vacuole fusion. Natamycin shows low MIC against the dematiaceous fungi, Curvularia species.

Formula: C₃₃H₄₇NO₁₃
MW: 665.733 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00135085
CAS Number: 7681-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR