Flame Resistant/ARC Rated High Visibility Class 3 Mesh Vest, Lakeland Industries
Supplier: Lakeland
V8AM0123ZL-MD V8AM0123ZL-LG V8AM0123ZL-XL V8AM0123ZL-2X V8AM0123ZL-3X V8AM0123ZL-4X V8AM0123ZL-5X
75923-620EA 206.96 USD
75923-620 75923-622 75923-624 75923-626 75923-628 75923-630 75923-632
Flame Resistant/ARC Rated High Visibility Class 3 Mesh Vest, Lakeland Industries
Workwear
High Visibility ANSI Class 3 rated vest of Flame Resistant/ARC rated modacrylic mesh fabric.
  • FR/ARC rated Modacrylic Mesh fabric
  • Non-conductive zipper closure
This ANSI 107-Class 3 Vest features a non-conductive zipper front closure, 3 M Scotchlite reflective material, and carries an ATPV value of 4.6 Cal/cm2. The Modacrylic Mesh fabric will never lose its FR and ARC rated protective properties.
Certifications: ANSI 107-Class 3. ASTM F1506.