Cryogenic Transfer Hoses, Western Enterprises
Supplier: Western Enterprises
312-WMH-2-11 312-WMH-2-13 312-WMH-2-10 312-WMH-2-9 312-WMH-2-12
300067-118EA 994.64 USD
300067-118 300067-112 300067-116 300067-126 300067-114
Cryogenic Transfer Hoses, Western Enterprises
Welding Supplies
Use in cyro-biological and medical systems and gas filling plants.
  • Cleaned for oxygen service
  • Exceptional flexibility, hose resists kinking
  • Machined CGA stainless steel end connections
  • Constructed of 316 stainless steel for all wetted parts