Benzene-[D₆] (99.5% D) (Isotopic)

Supplier: CAMBRIDGE ISOTOPE LABS INC MS
Danger

Synonyms: Benzene-[²H₆]

DLM-1-10X1 DLM-1-25 DLM-1-10X0.6 DLM-1-100 DLM-1-5 DLM-1-10X0.75 DLM-1-50 DLM-1-10
101322-134EA 109.85 USD
101322-134 101322-108 101322-128 101322-106 101322-110 101322-132 101322-112 101322-104
Benzene-[D₆] (99.5% D) (Isotopic)
Benzene-[D6]
Formula: C₆D₆
MW: 84.07 g/mol
MDL Number: MFCD00003010
CAS Number: 1076-43-3
UN: 1114
ADR: 3,II

Order Now

Specification Test Results

Purity (Isotopic Assay) 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR