Polyethylene glycol monomethyl ether 50%

Supplier: Rigaku Reagents

Synonyms: Methoxypolyethylene glycols, Methoxy poly(ethylene glycol), PEG MME, Poly(ethylene glycol) methyl ether, mPEG, Methoxypolyethylene glycols-average M.W. 1000, m-PEG-OH

1008067 1008066
101443-852EA 138.62 USD
101443-852 101443-460
Polyethylene glycol monomethyl ether 50%
Polyethylene glycol monomethyl ether

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: CH₃(OCH₂CH₂)nOH
MDL Number: MFCD00084416
CAS Number: 9004-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR