Polyethylene glycol 8,000 50% (w/v)
Supplier: Rigaku Reagents
Synonyms:
PEG 8,000, Ethylene glycol polymer 8000, PEG 8000
1008062
1008063
101443-486EA
103.31
USD
101443-486
101443-878
Polyethylene glycol 8,000 50% (w/v)
Polyethylene glycol 8,000
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.
Formula:
H(OCH₂CH₂)nOH MW: 8000 g/mol Boiling Pt: 250 |
MDL Number:
MFCD01779615 CAS Number: 25322-68-3 Merck Index: 12,07729 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...