Potassium dihydrogen phosphate 1.5 M
Supplier: Rigaku Reagents
Synonyms:
Monopotassiumphosphate, Potassium orthophosphate monobasic, prim.-Potassium phosphate, Potassium dihydrogenorthophosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate
1008080
1008081
101443-320EA
75.85
USD
101443-320
101443-668
Potassium dihydrogen phosphate 1.5 M
Potassium dihydrogen phosphate
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.
Formula:
KH₂PO₄ MW: 136.09 g/mol |
MDL Number:
MFCD00011401 CAS Number: 7778-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...