(±)-2-Methyl-2,4-pentanediol ≥99% (v/v)

Supplier: Rigaku Reagents

Synonyms: Hexylene glycol, (±)-2,4-Dihydroxy-2-methyl pentane, (±)-2,4-Dihydroxy-2-methyl pentane, 2-Methylpentane-2,4-diol, Hexylene Glycol

1008408 1008409
101443-382EA 116.38 USD
101443-382 101443-740
(±)-2-Methyl-2,4-pentanediol ≥99% (v/v)
(±)-2-Methyl-2,4-pentanediol

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: C₆H₁₄O₂
MW: 118.18 g/mol
MDL Number: MFCD00004547
CAS Number: 107-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR