(±)-1,2-Propanediol ≥99.5%

Supplier: Rigaku Reagents

Synonyms: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane

1008427 1008426
101443-896EA 128.15 USD
101443-896 101443-492
(±)-1,2-Propanediol ≥99.5%
Propylene glycol

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: CH₃CH(OH)CH₂OH
MW: 76.1 g/mol
Boiling Pt: 187.6
Melting Pt: -59
Flash Pt: 99
MDL Number: MFCD00064272
CAS Number: 57-55-6
Merck Index: 13,07947
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR