Glycerine ≥99.5% (v/v)

Supplier: Rigaku Reagents

1008402 1008403
101443-288EA 117.69 USD
101443-288 101443-624
Glycerine ≥99.5% (v/v)
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
MDL Number: MFCD00004722
CAS Number: 56-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR